Learn More
Kimberly-Clark™ Kimtech™ WetTask™ SXX Wipers
Kimtech™ WetTask SXX Wipers are ideal for solvent use, surface preparation and wiping harsh chemicals in industrial and healthcare sectors.
Brand: Kimberly-Clark™ 7769
159.02 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- 6 x 175 white sheets (1,050 total)
- Low-lint
- Silicone free
- pH stable
- Ideal for solvent use, surface preparation and wiping harsh chemicals
- Designed with the aviation transport sector
- Certified to requirements of BMS 15-5F and AMZ 3819B
Specifications
Light-duty Industrial Wiper | |
15.2 cm | |
22.5 cm | |
15.2 x 22.5 cm |
6 in. | |
8.9 in. | |
White | |
WetTask™ Roll Wiper Dispenser |