Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 04355814
This item is not returnable.
View return policy
Description
- With NS socket and cone
- With scale (approx. volume/mL)
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
500 mL | |
29/32 NS | |
Yes |
Borosilicate Glass | |
Pressure Equalizing Tube | |
Dropping Funnel |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug > Capacity: 500mL; Socket: 29/32 NS; Cone 29/32 NS
Spot an opportunity for improvement?Share a Content Correction