Learn More
Keystone Cleanroom Products 1073B Tyvek™ Autoclavable Stopper Bags
Keystone Cleanroom Products 1073B Tyvek™ Autoclavable Stopper Bags are breathable, and provide an effective bacterial barrier
Brand: Keystone Cleanroom Products SBMGTVKHS2026
This item is not returnable.
View return policy
Description
- Made from DuPont Tyvek and a polyethylene/HDPE film
- This combination of materials yields superior seal strength for stoppers, seals, and other critical components, or equipment parts
- These autoclavable bags are lint-free, moisture resistant, puncture resistant, and extremely tough
- Tyvek™ side offers high breathability while providing an effective bacterial barrier
- Clear film side offers excellent clarity combined with strength and durability
- Can be used in gas or steam sterilizers; and can also be irradiated
- Bags should be heat sealed closed, using a validate-able impulse, or continuous band sealer
Specifications
1073B Tyvek™ | |
Certificate of Conformance (COC) | |
Steam Sterilization | |
500/Pk. |
Autoclavable | |
White | |
HDPE | |
20 x 26 in. |