Learn More
Viking Optical™ 10X Folding Magnifier
Brand:  Viking Optical™ 5009
Encompass_Preferred
 View more versions of this product
Product Code. 12896325
  • 46.43 € / Each
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Pocket doublet type, giving tolerably flat field with minimum chromatic aberration. Lens approximately 19mm diameter. Mounted in metal frame.
Specifications
Specifications
11 mm