missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cultrex Spheroid Formation Extracellular Matrix, 10X

Supports spheroid formation for human breast cancer cells (MDA-MB-231) over a 72 hour period.

Brand:  R&D Systems™ 3500-096-01

261.77 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18711118

  • 333.00 € / 600µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Source: Murine Engelbreth-Holm-Swarm (EHS) tumor.
  • Storage Buffer: Dulbecco’s Modified Eagle’s medium (DMEM) containing 10 μg/mL gentamicin sulfate
  • Storage/Stability: Product is stable for a minimum of 3 months from date of shipment. Store at –80°C; keep frozen. Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity
Specifications

Specifications

Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Mouse
Basement Membrane Extracts
600 μL
Murine Engelbreth-Holm-Swarm (EHS) tumor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers