Learn More
11-Azidoundecyltrimethoxysilane, 94%, Thermo Scientific Chemicals
107.00 € - 290.00 €
Chemical Identifiers
CAS | 334521-23-2 |
---|---|
Molecular Formula | C14H31N3O3Si |
Molecular Weight (g/mol) | 317.51 |
MDL Number | MFCD11982878 |
InChI Key | ACSJXAKDVNDMME-UHFFFAOYSA-N |
Synonym | 11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide |
PubChem CID | 46779110 |
SMILES | CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15467237
|
Thermo Scientific Alfa Aesar
H34298.MD |
250 mg |
107.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15457237
|
Thermo Scientific Alfa Aesar
H34298.03 |
1 g |
290.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
334521-23-2 | |
317.51 | |
ACSJXAKDVNDMME-UHFFFAOYSA-N | |
46779110 |
C14H31N3O3Si | |
MFCD11982878 | |
11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide | |
CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC |
Specifications
100°C to 105°C (0.04mmHg) | |
C14H31N3O3Si | |
MFCD11982878 | |
ACSJXAKDVNDMME-UHFFFAOYSA-N | |
(11-azidoundecyl)trimethoxysilane | |
46779110 | |
94% | |
11-Azidoundecyltrimethoxysilane |
334521-23-2 | |
N3(CH2)11Si(OCH3)3 | |
11-azidoundecyl trimethoxysilane, 11-trimethoxysilyl undecyl azide | |
CO[Si](CCCCCCCCCCCN=[N+]=[N-])(OC)OC | |
317.51 | |
317.51 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only