Learn More
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide, 95%, Thermo Scientific Chemicals
116.58 € - 381.90 €
Chemical Identifiers
CAS | 207399-07-3 |
---|---|
Molecular Formula | C36H44ClIN2 |
Molecular Weight (g/mol) | 667.116 |
MDL Number | MFCD00191907 |
InChI Key | IRPKBYJYVJOQHQ-UHFFFAOYSA-M |
Synonym | 2-2-2-chloro-3-1,3-dihydro-3,3-dimethyl-1-propyl-2h-indol-2-ylidene ethylidene-1-cyclohexen-1-yl ethenyl-3,3-dimethyl-1-propylindolium iodide |
PubChem CID | 118987185 |
IUPAC Name | 2-[(2E)-2-[2-chloro-3-[(E)-2-(3,3-dimethyl-1-propylindol-1-ium-2-yl)ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethyl-1-propylindole;iodide |
SMILES | CCCN1C2=CC=CC=C2C(C1=CC=C3CCCC(=C3Cl)C=CC4=[N+](C5=CC=CC=C5C4(C)C)CCC)(C)C.[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15495135
|
Thermo Scientific Alfa Aesar
H32150.03 |
1 g |
116.58 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405145
|
Thermo Scientific Alfa Aesar
H32150.06 |
5 g |
381.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications11-Chloro-1,1′-di-n-propyl-3,3,3′,3′-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide is used as a laser and IR dye. It is also used as lithographic computer to plate laser applications.
Solubility
It is soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
207399-07-3 | |
667.116 | |
IRPKBYJYVJOQHQ-UHFFFAOYSA-M | |
118987185 | |
CCCN1C2=CC=CC=C2C(C1=CC=C3CCCC(=C3Cl)C=CC4=[N+](C5=CC=CC=C5C4(C)C)CCC)(C)C.[I-] |
C36H44ClIN2 | |
MFCD00191907 | |
2-2-2-chloro-3-1,3-dihydro-3,3-dimethyl-1-propyl-2h-indol-2-ylidene ethylidene-1-cyclohexen-1-yl ethenyl-3,3-dimethyl-1-propylindolium iodide | |
2-[(2E)-2-[2-chloro-3-[(E)-2-(3,3-dimethyl-1-propylindol-1-ium-2-yl)ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethyl-1-propylindole;iodide |
Specifications
207399-07-3 | |
C36H44ClIN2 | |
2-2-2-chloro-3-1,3-dihydro-3,3-dimethyl-1-propyl-2h-indol-2-ylidene ethylidene-1-cyclohexen-1-yl ethenyl-3,3-dimethyl-1-propylindolium iodide | |
IRPKBYJYVJOQHQ-UHFFFAOYSA-M | |
2-[(2E)-2-[2-chloro-3-[(E)-2-(3,3-dimethyl-1-propylindol-1-ium-2-yl)ethenyl]cyclohex-2-en-1-ylidene]ethylidene]-3,3-dimethyl-1-propylindole;iodide | |
118987185 | |
95% |
232°C to 234°C | |
MFCD00191907 | |
It is soluble in methanol. | |
CCCN1C2=CC=CC=C2C(C1=CC=C3CCCC(=C3Cl)C=CC4=[N+](C5=CC=CC=C5C4(C)C)CCC)(C)C.[I-] | |
667.116 | |
667.12 | |
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only