missing translation for 'onlineSavingsMsg'
Learn More

11-(Pentafluorophenoxy)undecyltriethoxysilane, 95%, Thermo Scientific™

Product Code. 15425847
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197981-13-7
Molecular Formula C23H37F5O4Si
Molecular Weight (g/mol) 500.622
MDL Number MFCD13181914
InChI Key YAAQJDPGHMLLJH-UHFFFAOYSA-N
Synonym 11-pentafluorophenoxyundecyl triethoxysilane, 11-pentafluorophenoxy undecyltriethoxysilane, triethoxy 11-2,3,4,5,6-pentafluorophenoxy undecyl silane
PubChem CID 99738222
IUPAC Name triethoxy-[11-(2,3,4,5,6-pentafluorophenoxy)undecyl]silane
SMILES CCO[Si](CCCCCCCCCCCOC1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC

Specifications

Boiling Point 150°C to 155°C (0.04mmHg)
Quantity 1 g
Formula Weight 500.62
Percent Purity 95%
Refractive Index 1.4340-1.4380
Sensitivity Moisture sensitive
Chemical Name or Material 11-(Pentafluorophenoxy)undecyltriethoxysilane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'