Learn More
Hanna Instruments™ Chemical Test Kit Replacement Reagents
Pre-made reagents prepared with high-purity chemicals for ease of use. Hanna Instruments™ Chemical Test Kit Replacement Reagents are pre-measured for fast and accurate titrations using the appropriate Hanna Chemical Test Kit.
38.95 € - 221.00 €
Product Code | Brand | For Use With (Application) | No. of Tests | Test Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | No. of Tests | Test Method | Price | Quantity & Availability | |||
11682350
|
Hanna Instruments™
HI3810100 |
Dissolved Oxygen determination | 100 | Titration |
221.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11652390
|
Hanna Instruments™
HI3835100 |
Salinity determination | 110 | Titration |
161.00 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
11602360
|
Hanna Instruments™
HI3811100 |
Alkalinity determination | 110 | Titration |
200.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11612360
|
Hanna Instruments™
HI3812100 |
Total hardness determination | 100 | Titration |
140.00 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
11652360
|
Hanna Instruments™
HI3815100 |
Chloride determination | 110 | Titration |
216.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11692360
|
Hanna Instruments™
HI3818100 |
Carbon Dioxide determination | 110 | Titration |
202.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11622370
|
Hanna Instruments™
HI3820100 |
Acidity determination | 100 | Titration |
208.00 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
11662380
|
Hanna Instruments™
HI3831F050 |
Free Chlorine determination | 50 | Titration |
40.30 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11682380
|
Hanna Instruments™
HI3831T050 |
Total Chlorine determination | 50 | Titration |
38.95 €
Each |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||
10029633
|
Hanna Instruments™
|
Phosphate determination | 50 | Titration |
57.55 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11632390
|
Hanna Instruments™
HI3834050 |
Iron determination | 50 | Titration |
125.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
These high quality reagents are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability. Replacement titration reagent sets provide significant cost savings compared to purchasing an entirely new Test Kit.
Chemical Test Kit Replacement Reagents are available for the following Chemical Test Kits:
- Acidity Test Kit
- Alkalinity Test Kit
- Ascorbic Acid Test Kit
- Cardon Dioxide Test Kit
- Chloride Test Kit
- Dissolved Oxgen Test Kit
- Free and Total Chlorine (Low, Medium, High Range) Test Kit
- Free Chlorine Test Kit
- Total Chlorine Test Kit
- Iron (Medium Range) Test Kit
- Phosphate Test Kit
- Salinity Test Kit
- Sulfite Test Kit
- Total Hardness Test Kit
Specifications
1 Set |