Learn More
Cole-Parmer™ 1104-1 Alconox Cleaner
Cole-Parmer 1104-1 Alconox Cleaner, For Use With: For Manual or Ultrasonic Cleaning, Certifications/Compliance: FDA compliant, Quantity: 1Each
Brand: Cole-Parmer™ 17775-05
65.36 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Cole-Parmer 1104-1 Alconox Cleaner, For Use With: For Manual or Ultrasonic Cleaning, Certifications/Compliance: FDA compliant, Quantity: 1Each
Specifications
Alconox Cleaner | |
FDA compliant | |
For Manual or Ultrasonic Cleaning | |
1 Ea. |
Safety and Handling
- Alconox 1104-1
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P321-Specific treatment (see on this label).
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.