missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 11mm Push On Cap, for 11mm Crimp Vials, thinned penetration area

Polyethylene caps are chemically inert and suitable for most chromatography applications.

Brand:  Fisherbrand™ 11 08 1676

73.09 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 11836201

  • 77.15 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 11mm PE Push On Cap
  • Blue
  • With thinned penetration area
  • Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
Polyethylene
Push On Cap
11 mm
100
ND11
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ 11mm Push On Cap, for 11mm Crimp Vials, thinned penetration area > PE, closed top,blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.