Learn More
Labconco™ 12-Port Drying Chamber for FreeZone™ Freeze-Dry Systems; PTFE-coated
Drying Chamber with corrosion-resistant interior for use on FreeZone™ Freeze Dryers.
Brand: Labconco™ 7522801
This item is not returnable.
View return policy
Description
12-Port Chamber, PTFE- 12-port drying chamber made of type 304 stainless steel with corrosion-resistant PTFE-coated interior
- Includes arcylic lid with neoprene gasket, latching pin, and 12 neoprene valves with molded plastic knobs for 1/2 in. & 3/4 in. adapters
- For use with FreeZone™ 1, 2.5, 4.5, 6, 8, 12, or 18 Liter Freeze Dryers
Specifications
PTFE-coated Stainless Steel | |
To provide a means to attach flasks to freeze dry system or contain samples in bulk | |
8 in. | |
10 lb. |
12 | |
FreeZone™ Freeze Dryers | |
9.75 in. |