Learn More
Fisherbrand™ 12-Volt Sprout™ Power Adapter
An adapter for the Sprout Mini Centrifuge.
Brand:  Fisherbrand™ 120570
Product Code. 15827944
  • 28.80 € / Each
Explore more special offers
Description
Description
  • 12 Volt
  • Enables a convenient way to spin down and prepare samples outside of a conventional laboratory environment
Specifications
Specifications
Sprout™ Mini Centrifuge
12V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ 12-Volt Sprout™ Power Adapter > 12 Volt