Learn More
BRAND™ 12-well Insert 2in1, PET-Membrane
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
Brand:  BRAND™ 782730
369.30 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 16184957
  • 422.00 € / Pack of 48
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Use Inserts in a hanging or standing position
  • For 12-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12-well Insert 2in1, PET-Membrane 0.4 μm
0.4 μm
Sterile
PET
48 x 1
12