missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 13mm Clear Glass Crimp Top Vials, Neisseria Meningitidis, Polyvalent A-D

For use with snap caps or aluminum crimp seal closures, these high-quality 13mm crimp/snap top vials are 4mL, 15x45mm vials and fit a variety of autosamplers.

Brand:  Thermo Scientific™ C4015-4

24.76 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.


Product Code. 11931523

  • 26.17 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, Thermo Scientific™ National™ 13mm Snap/Crimp Vials features a 13mm crimp/snap-ring finish for use with 13mm Aluminum Seals or Kim-Snap Closures. Polyspring inserts are self-aligning and provide a cushion against needle contact.

Autosampler compatibility*

Dionex™

Shimadzu™

Spark Holland™

Varian™

VWR™ (Merck)/Hitachi™

Waters™ (Wisp 48 Position Carousel)

*See the Columns and Consumables Catalog for list of autosampler compatibility.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neisseria Meningitidis; Polyvalent A-D
15 mm
15 mm
4.8 mL
Flat Bottom
Crimp/Snap
100
13 mm
4.25 mL
13 mm
Clear
Glass
<800 μL
4.8 mL
45 mm
13 mm Crimp/Snap Vial
No
Aluminum Seals or Kim-Snap™ Closures
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.