missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ 140 μL Na-hep Capillaries

An excellent choice for blood gas analysis.

Brand:  Hirschmann™ 9222275

Product Code. 10233531

  • 163.00 € / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sodium heparinized
  • With LOT number on packaging
  • Outer diameter of 2.35 mm, inner diameter of 1.55 mm
  • Labelled with: usable until, CE pursuant to IVD 98/EC
Specifications

Specifications

Na-hep Capillary
75 mm
140 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.