Learn More
Keystone Cleanroom Products 1421B Tyvek™ Autoclave Sterilization Wrapper
GREENER_CHOICE
Autoclavable Wrapper provides an excellent bacterial and microbial barrier for superior performance post steam sterilization.
Brand:  Keystone Cleanroom Products 70TK5454
 View more versions of this product
Product Code. 30020285
  • 1750.00 € / Pack of 125
Explore more special offers
Description
Description
Tyvek™ Autoclavable Wrappers are breathable while providing an excellent bacterial and microbial barrier for superior performance post steam sterilization.
  • Lint-free (Helmke Drum) means no airborne particulates are generated during its use as sterilization packaging
  • Clean and consistently cut using an automated CAD system per ISO 8 Requirements
  • Packaged in an ISO 6 Controlled Clean Room
  • Designed for Steam Sterilization
  • Hydrophobic
  • Resists tearing and punctures
Specifications
Specifications
Sterilization Wrap
Certificate of Conformance (COC)
White
Steam Sterilization
Tyvek
Up to 250°F
54 x 54 in.
137 cm
137 cm
Autoclavable
None
Single-use
Pharmaceutical Equipment and Components
Non-sterile
Up to 125°C
54 in.
54 in.