missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ 150mm Correlated Flowmeters with Valve

Brand:  Masterflex™ 03294-14

 View more versions of this product

Product Code. 15389482

  • 297.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Variable Area Flowmeter
814 mL/min.
200 psig
728 mL/min. CO2, 753 mL/min. O2, 824 mL/min. N2, 676 mL/min. Ar, 2496 mL/min. H2
150 mm
121°C
±2% (Full Scale)
Stainless Steel (316)
1/8 in. NPT (F)
Perfect for applications demanding high resolution
±0.25%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.