Learn More
Corning™ 1536-Well Polystyrene NBS™-Treated Microplates
GREENER_CHOICE
Proprietary nonbinding surface is ideal for homogeneous assays with sticky enzymes or for reducing nonspecific binding to microplate wells
Brand:  Corning™ 3728
 View more versions of this product
Product Code. 10027322
  • 1363.53 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Order Info
Packaged in bags of ten
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
32 x 48
Black
Polystyrene
8 μL
12.5 μL
Without barcode
10/Pk.
Black
Without Lid
5 inner packs of 10
1536-Well Cell Culture Multiwell Plates
1536
Non-sterile
Flat Bottom (F-well)
50/Cs.
50/Cs.
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Product Certifications
Offerte speciali
Offerte speciali