missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tuttnauer 160L Benchtop Autoclave
The 5075EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Brand: Tuttnauer 5075EL-380-D
This item is not returnable.
View return policy
Description
Designed for your laboratory applications. The 5075EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
- Advanced Control System - Take Advantage of Tuttnauer’s state-of-the-art control system with multi-color display
- Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
- Optional Internal Printing - Documents date, time, temperature and pressure
- Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
- High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
- Diverse Load Types - The 5075EL-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications
35.2 gal. | |
75 cm | |
30 in. | |
44.8 in. | |
Benchtop Autoclave | |
105°C to 137°C | |
316L Stainless Steel |
160 L | |
20 in. | |
50 cm | |
112 cm | |
221°F to 279°F | |
Three Phase | |
Non-sterile |