Learn More
Fisherbrand™ Plastic Lock Bacti Caps
Made from chemically resistant virgin plastic, Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange. They are ideal for use in science, research, and other laboratories.
Brand:  Fisherbrand™ BC16GREEN
Product Code. 11517153
  • 139.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs
  • Suitable for all standard test tube sizes, they fit rimless and plastic tubes with an outside diameter of 13mm, 16mm, 19mm, 25mm and 38mm
  • The three flexible fins molded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur. Bacti-Caps™ are made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters
  • Improved culture growth
  • Color coded
  • Controlled gas interchange
Order Info
dimClosureColor
Specifications
Specifications
Lock Bacti Cap
Plastic
100 Pack
Test Tubes
Autoclavable
Green
16 mm