missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Chemical Spoon w/ Spatula Ends

Manufactured from stainless steel. Bochem™ 18/10 Stainless Steel Chemical Spoon w/ Spatula Ends is multi-purpose.

Brand:  Bochem™ 3425

 View more versions of this product

Product Code. 10602961

  • 8.70 € / Each
Estimated Shipment: 09-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spatula handle
  • Autoclavable
  • Stainless steel
Specifications

Specifications

1 Ea.
Chemical Scoop
50 mm
Stainless Steel (18/10)
300 x 37 mm
Flat
Autoclavable
Multi-purpose spoon for a wide range of uses
37 mm
2mm
Stainless Steel (18/10)
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers