Learn More
Bochem™ 18/10 Stainless Steel Dissecting Needle
For use with Bochem needle holder Kolle 12000. Bochem™ 18/10 Stainless Steel Dissecting Needle is manufactured from stainless steel.
Brand:  Bochem™ 12005
 View more versions of this product
Product Code. 11835091
  • 27.50 € / Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Type 1: eye
  • Type 2: needle
  • Type 3: lancet
  • For use with Bochem needle holder Kolle 12000
  • Manufactured from stainless steel
Specifications
Specifications
100 mm
Lancet
10/pk.
Stainless Steel (18/10)
Type 3 Lancet Holder KOLLE 12000