missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Dissecting Needle

For use with Bochem needle holder Kolle 12000. Bochem™ 18/10 Stainless Steel Dissecting Needle is manufactured from stainless steel.

Brand:  Bochem™ 12001

 View more versions of this product

Product Code. 11845101

This item is not returnable. View return policy

Description

Description

  • Type 1: eye
  • Type 2: needle
  • Type 3: lancet
  • For use with Bochem needle holder Kolle 12000
  • Manufactured from stainless steel
Specifications

Specifications

1.5 mm
Stainless Steel (18/10)
Type 1 Eye Holder KOLLE 12000
100 mm
Eye
10/pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.