Learn More
Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas
Manufactured from stainless steel. Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas are multi-purpose.
Brand:  Bochem™ 3016
 View more versions of this product
Product Code. 10060841
  • 2.45 € / Each
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Double endet
  • Stainless steel
  • Autoclavable
  • Straight
Specifications
Specifications
1 Ea.
Straight
Multi-purpose spatula for a wide range of uses
4 mm
Stainless Steel (18/10)
100 x 4 mm
100 mm
Autoclavable
Double Ended Micro Spatula
35 mm
4 mm
2mm
Stainless Steel (18/10)