Learn More
Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas
Manufactured from stainless steel. Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas are multi-purpose.
Brand:  Bochem™ 3027
 View more versions of this product
Product Code. 10259070
  • 2.75 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Double endet
  • Stainless steel
  • Autoclavable
  • Straight
Specifications
Specifications
1 Ea.
Straight
Multi-purpose spatula for a wide range of uses
6 mm
Stainless Steel (18/10)
210 x 6 mm
210 mm
Autoclavable
Double Ended Micro Spatula
55 mm
6 mm
2mm
Stainless Steel (18/10)