missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Micro Scoops, Half-rounded
Add Bochem™ 8/10 Stainless Steel Micro Scoops, Half-rounded to your arsenal of laboratory instruments.
Brand: Bochem™ 3304
This item is not returnable.
View return policy
Description
- Type 1
- Half-rounded
- Also available in wide scoop
- 18/10 stainless steel
Specifications
1 Ea. | |
Half-rounded | |
50 mm | |
4 mm | |
2mm | |
Stainless Steel (18/10) |
Autoclavable | |
Micro Scoop | |
4 mm | |
Stainless Steel (18/10) | |
185 x 4 mm | |
185 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction