missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Poly Spoons

Manufactured from 18/10 stainless steel

Brand:  Bochem™ 3404

 View more versions of this product

Product Code. 10506562

  • 7.45 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spoon comes in three different types
  • Autoclavable
  • Made of 18/10 stainless steel
  • Multi-purpose spoon for a wide range of uses
Specifications

Specifications

1 Ea.
Poly Spoon
30 mm
Stainless Steel (18/10)
300 x 13 mm
300 mm
Autoclavable
Multi-purpose spoon for a wide range of uses
13 mm
5mm
Stainless Steel (18/10)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.