missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Spatulas, Spoon/round/bent
Manufactured from 18/10 stainless steel
Brand: Bochem™ 3203
This item is not returnable.
View return policy
Description
- Made from 18/10 stainless steel
Specifications
1 Ea. | |
Spoon/Round/Bent | |
Used for a variety of applications | |
3mm | |
180 mm |
Autoclavable | |
Spatula | |
Stainless Steel (18/10) | |
Stainless Steel (18/10) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction