Learn More
Bochem™ 18/10 Stainless Steel Spoon Spatulas
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Spoon Spatulas serve a wide variety of functions in the lab.
Brand:  Bochem™ 3473
 View more versions of this product
Product Code. 11875071
  • 85.80 € / Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from 18/10 stainless steel
Specifications
Specifications
10 Pk.
50 mm
10 mm
210 mm
Spoon Spatula
12 mm
Stainless Steel (18/10)