missing translation for 'onlineSavingsMsg'
Learn More

18-beta-Glycyrrhetinic acid, 98+%

Product Code. 10020960 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10020960

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 471-53-4 | C30H46O4 | 470.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-53-4
Molecular Formula C30H46O4
Molecular Weight (g/mol) 470.69
MDL Number MFCD00003706,MFCD00066716
InChI Key MPDGHEJMBKOTSU-YKLVYJNSSA-N
Synonym enoxolone, glycyrrhetinic acid, uralenic acid, glycyrrhetic acid, 18-beta-glycyrrhetinic acid, glycyrrhetin, arthrodont, jintan, 18beta-glycyrrhetinic acid, rhetinic acid
PubChem CID 10114
ChEBI CHEBI:30853
IUPAC Name (2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid
SMILES CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 292°C to 295°C
Color White
Loss on Drying 1% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 3644
Specific Rotation Condition +165° (20°C c=1,CHC3,on dry ba)
Specific Rotation + 165
Formula Weight 470.69
Percent Purity 98+%
Related Substances 2% max. (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 18-β-Glycyrrhetinic acid, 98+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.