Learn More
Fisherbrand™ 18-Place Slide Staining Tray Set, Black
Slide tray that is ideal for clinical and research laboratory staining, rinsing, and drying applications.
Brand:  Fisherbrand™ 120879F
131.59 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Product Code. 17784749
  • 156.00 € / Each
Explore more special offers
Description
Description
  • Durably constructed from solvent resistant polypropylene blend
  • Non-skid silicone feet keep tray in place
  • Two ways to empty stained liquid
  • Equipped with handle cut outs for easy handling
  • Humidity retention wells for specialized staining needs
Specifications
Specifications
Slide Staining Tray
Microscope slide staining, rinsing and drying applications
1 black base and tray, 1 black cover, and 2 drain plugs
18
Standard microscope slides (76 x 26 mm)
1 Ea.