Learn More
Tocris™ 1A-116
Rac1 inhibitor; blocks Rac1-P-Rex1 interaction
Brand:  Tocris™ 6701/10
215.14 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18054522
  • 266.00 € / 10mg
Explore more special offers
Description
Description
1A-116 is a Rac1 inhibitor. Reduces intracellular Rac1-GTP levels and blocks Rac1-P-Rex1 interaction in vitro. Exhibits no effect on GTPase activity of closely related Cdc42. Displays antiproliferative, anti-invasive and pro-apoptotic activity in several cancer cell lines. Exhibits antimetastatic activity in vivo. Also restores antiproliferative effects of Tamoxifen (Cat. No. 6342) in resistant breast cancer cell.
Specifications
Specifications
N-(3,5-Dimethylphenyl)-N-[2-(trifluoromethyl)phenyl]guanidine
10 mg
307.31
98%
1430208-73-3
C16H16F3N3
Inhibitor