missing translation for 'onlineSavingsMsg'
Learn More

1alpha, 25-Dihydroxy VD2-d6, MedChemExpress

MedChemExpress 1alpha, 25-Dihydroxy VD2-D6 is a deuterated form of vitamin D.

POA - 3249.45 €

Specifications

Chemical Name or Material 1alpha, 25-Dihydroxy VD2-d6
CAS 216244-04-1
Purity Grade Notes Research
Molecular Formula C28H38D6O3
SMILES O[C@H](C/1=C)C[C@H](O)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16491499
Encompass_Preferred
View Documents
Medchem Express
HY-15327
1 mg
3249.45 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16429047
Encompass
View Documents
Medchem Express
HY-15327
5 mg
POA
5mg
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

1alpha, 25-Dihydroxy VD2-D6 is a deuterated form of vitamin D.
Specifications

Specifications

1alpha, 25-Dihydroxy VD2-d6
Research
O[C@H](C/1=C)C[C@H](O)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2
434.68
Metabolism-sugar/lipid metabolism
216244-04-1
C28H38D6O3
434.68
Research
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Please store the product under the recommended conditions in the Certificate of Analysis.

Recommended Storage : Please store the product under the recommended conditions in the Certificate of Analysis.

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.