missing translation for 'onlineSavingsMsg'
Learn More
1H-1,2,4-triazole-3-thiol, Thermo Scientific™ Product Code.: 10473014

1H-1,2,4-triazole-3-thiol, Thermo Scientific™

Product Code. 10473014
25g
Click to view available options
25g
This item is not returnable. View return policy

Product Code. 10473014

Brand: Thermo Scientific Maybridge SB00858EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3179-31-5
Assay Percent Range 97%
Molecular Formula C2H3N3S
MDL Number MFCD00005229
Quantity 25g
Synonym 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol
InChI Key AFBBKYQYNPNMAT-UHFFFAOYSA-N
SMILES S=C1NNC=N1
IUPAC Name 2,3-dihydro-1H-1,2,4-triazole-3-thione
Molecular Weight (g/mol) 101.13
PubChem CID 2723802
Formula Weight 101.13
Percent Purity 97%
Chemical Name or Material 1H-1, 2, 4-Triazole-3-thiol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-1,2,4-triazole-3-thiol, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.