missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluoro-1-decanol, 97%, Thermo Scientific™

Product Code. 11409946
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a chemical applied to the formation of a barrier and self-healing coating on zinc metal materials. It induces cell death and formation of oxidative or reactive oxygen species in cerebellar granule cells. It is used to make surfactant and polymeric products with a range of commercial applications (may contain small amounts of residual unreacted material). The polymers are primarily used for coating textiles, paper, and carpet for oil, stain, and water repellency.

Solubility
Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 678-39-7
Molecular Formula C10H5F17O
Molecular Weight (g/mol) 464.122
MDL Number MFCD00039544
InChI Key JJUBFBTUBACDHW-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluoro-1-decanol, 1h,1h,2h,2h-perfluorodecanol, 2-perfluoro-n-octyl ethanol, 1,1,2,2-tetrahydroperfluoro-1-decanol, 2-perfluorooctyl ethanol, 8:2 fluorotelomer alcohol, 1h,1h,2h,2h-perfluorodecan-1-ol, 8:2 ftoh, unii-um7l66ofe1, 1-decanol, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro
PubChem CID 69619
ChEBI CHEBI:63919
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecan-1-ol
SMILES C(CO)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 45°C to 50°C
Boiling Point 112°C to 114°C (10 mmHg)
Quantity 50 g
Beilstein 2227487
Solubility Information Soluble in chloroform and methanol.
Formula Weight 464.12
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-decanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'