missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyldimethylchlorosilane, tech. 90%, Thermo Scientific™

26.00 € - 390.00 €

Chemical Identifiers

CAS 74612-30-9
Molecular Formula C12H10ClF17Si
Molecular Weight (g/mol) 540.72
MDL Number MFCD00042343
InChI Key JHCJWHBMXWOYDE-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyldimethylchlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl dimethylchlorosilane, 2-heptadecafluorooctyl ethylchlorodimethylsilane, chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethylsilane, chloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-dimethylsilane, chlorodimethyl-3,3,4,4,5,5,6,6,7,7,8,8, 9,9,10,10,10-heptadecafluorodecylsilane, silane,chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethyl
PubChem CID 144665
IUPAC Name chloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-dimethylsilane
SMILES C[Si](C)(CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339245
View Documents
Thermo Scientific Alfa Aesar
L16582.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11349245
View Documents
Thermo Scientific Alfa Aesar
L16582.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11359245
View Documents
Thermo Scientific Alfa Aesar
L16582.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

74612-30-9
540.72
JHCJWHBMXWOYDE-UHFFFAOYSA-N
144665
C[Si](C)(CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)Cl
C12H10ClF17Si
MFCD00042343
1h,1h,2h,2h-perfluorodecyldimethylchlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl dimethylchlorosilane, 2-heptadecafluorooctyl ethylchlorodimethylsilane, chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethylsilane, chloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-dimethylsilane, chlorodimethyl-3,3,4,4,5,5,6,6,7,7,8,8, 9,9,10,10,10-heptadecafluorodecylsilane, silane,chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethyl
chloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-dimethylsilane

Specifications

197°C to 198°C
C12H10ClF17Si
UN2987
1h,1h,2h,2h-perfluorodecyldimethylchlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl dimethylchlorosilane, 2-heptadecafluorooctyl ethylchlorodimethylsilane, chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethylsilane, chloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-dimethylsilane, chlorodimethyl-3,3,4,4,5,5,6,6,7,7,8,8, 9,9,10,10,10-heptadecafluorodecylsilane, silane,chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethyl
C[Si](C)(CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)Cl
540.72
540.73
Technical
74612-30-9
MFCD00042343
4584072
JHCJWHBMXWOYDE-UHFFFAOYSA-N
chloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-dimethylsilane
144665
90%
1H,1H,2H,2H-Perfluorodecyldimethylchlorosilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.