missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Perfluorodecyltrichlorosilane, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1H,1H,2H,2H-Perfluorodecyltrichlorosilane is a halogenated compound used for biochemical research. It is applied to movable microparts of microelectromechanical systems , thereby reducing the surface energy and preventing the stickiness. Further, it is used to coat micro- and nano-features on stamps for a nanoimprint lithography. In addition, it is used in the preparation of (1H,1H,2H,2H-heptadecafluorodecyl)trimethoxysilane by reacting with methanol.
Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.
Notes
Moisture sensitive. Incompatible with oxidizing agents, strong acids and bases.
1H,1H,2H,2H-Perfluorodecyltrichlorosilane is a halogenated compound used for biochemical research. It is applied to movable microparts of microelectromechanical systems , thereby reducing the surface energy and preventing the stickiness. Further, it is used to coat micro- and nano-features on stamps for a nanoimprint lithography. In addition, it is used in the preparation of (1H,1H,2H,2H-heptadecafluorodecyl)trimethoxysilane by reacting with methanol.
Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.
Notes
Moisture sensitive. Incompatible with oxidizing agents, strong acids and bases.
Specifications
Specifications
Boiling Point | 224°C |
Physical Form | Liquid |
Quantity | 1 g |
Linear Formula | CF3(CF2)7(CH2)2SiCl3 |
UN Number | UN2987 |
Beilstein | 5889653 |
Flash Point | >200°C (392°F) |
Solubility Information | Miscible with tetrahydrofuran,tetrhydropyran,toluene and other organic solvents. |
Formula Weight | 581.56 |
Percent Purity | 96% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1H,1H,2H,2H-Perfluorodecyltrichlorosilane, 96%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction