missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98%, Thermo Scientific Chemicals

30.35 € - 287.43 €

Chemical Identifiers

CAS 56602-33-6
Molecular Formula C12H22F6N6OP2
Molecular Weight (g/mol) 442.287
MDL Number MFCD00011948
InChI Key MGEVGECQZUIPSV-UHFFFAOYSA-N
Synonym bop reagent, 1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, castro's reagent, bop-reagent, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, ccris 2602, castros reagent, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
PubChem CID 151348
IUPAC Name benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
SMILES CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443187
View Documents
Thermo Scientific Alfa Aesar
A16140.03
1 g
30.35 €
1g
Please to purchase this item. Need a web account? Register with us today!
11453187
View Documents
Thermo Scientific Alfa Aesar
A16140.06
5 g
72.26 €
5g
Please to purchase this item. Need a web account? Register with us today!
11463187
View Documents
Thermo Scientific Alfa Aesar
A16140.14
25 g
287.43 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate is used as a reagent for peptide coupling, lactonization, selective esterification, amidation of alfa amino acids without racemization and synthesis of magnolamide for antioxidative activity and catalyst for 9-acridinecaroboxamide derivative. It is also used as a precursor for the synthesis of phenyl esters of amino acids. It acts as a substitute for (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate is used as a reagent for peptide coupling, lactonization, selective esterification, amidation of alfa amino acids without racemization and synthesis of magnolamide for antioxidative activity and catalyst for 9-acridinecaroboxamide derivative. It is also used as a precursor for the synthesis of phenyl esters of amino acids. It acts as a substitute for (Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) reagent.

Solubility
Soluble in methanol, acetone and dichloromethane. Insoluble in water.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.
Specifications

Chemical Identifiers

56602-33-6
442.287
MGEVGECQZUIPSV-UHFFFAOYSA-N
151348
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
C12H22F6N6OP2
MFCD00011948
bop reagent, 1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, castro's reagent, bop-reagent, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, ccris 2602, castros reagent, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate

Specifications

56602-33-6
C12H22F6N6OP2
UN1325
bop reagent, 1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, castro's reagent, bop-reagent, benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate, ccris 2602, castros reagent, tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate, benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
442.287
442.29
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate
∼138°C (decomposition)
MFCD00011948
4287025
MGEVGECQZUIPSV-UHFFFAOYSA-N
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
151348
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.