missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole, 99%

Product Code. 10429670 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10429670

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-14-7 | C6H5N3 | 119.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-14-7
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD00005699
InChI Key QRUDEWIWKLJBPS-UHFFFAOYSA-N
Synonym 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
PubChem CID 7220
ChEBI CHEBI:75331
IUPAC Name 2H-benzotriazole
SMILES C1=CC2=NNN=C2C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 94.0°C to 99.0°C
Color Beige-Yellow to White
Boiling Point 204.0°C (15.0 mmHg)
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 26, 38
Fieser 16,21
Merck Index 15, 1111
Solubility Information Solubility in water: 25g/L in water (20°C). Other solubilities: soluble in alcohol,benzene,toluene,chloroform,and dmf,soluble in ethanol,methanol and ethyleneglycol
Formula Weight 119.13
Percent Purity 99%
Physical Form Powder, Granules, Crystals, Needles or Flakes
Chemical Name or Material 1H-Benzotriazole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1H-Benzotriazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.