missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole-(ring-d4) solution, Honeywell Fluka™

10 μg/mL in acetone (15949681), 50 μg/mL in acetone (15900192) Analytical standard, reference material

307.00 €

Specifications

Packaging Glass Bottle
Grade Analytical Standard
Density 0.79 g/cm3
Boiling Point 55°C to 57°C
Flash Point -17°C
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15949681
View Documents
Honeywell Fluka
R1164-5ML
5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
Specifications

Specifications

Glass Bottle
0.79 g/cm3
-17°C
Liquid
Analytical Standard
55°C to 57°C
Parabenes
Videos
Safety and Handling

Safety and Handling

P210+P260+P280

H225+H319+H336

Danger

SDS
Documents

Documents

Product Certifications