missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazol-2-ylmethanol hydrochloride, 97%, Thermo Scientific™

Product Code. 10160632
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 116177-22-1
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.563
MDL Number MFCD03659700
InChI Key FEORNCWZOSTSRO-UHFFFAOYSA-N
Synonym 1h-imidazol-2-ylmethanol hydrochloride, 1h-imidazol-2-yl methanol hydrochloride, imidazolylmethanol hydrochloride, hydroxymethylimidazole hydrochloride, hydroxymethyl imidazole hydrochloride, imidazol-2-ylmethan-1-ol, chloride
PubChem CID 2776278
IUPAC Name 1H-imidazol-2-ylmethanol;hydrochloride
SMILES C1=CN=C(N1)CO.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 134.57
Percent Purity 97%
Chemical Name or Material 1H-imidazol-2-ylmethanol hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.