missing translation for 'onlineSavingsMsg'
Learn More
1H-Indazole-6-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 10636850

1H-Indazole-6-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 10636850
1 g, Glass bottle
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10636850

Brand: Thermo Scientific Acros 430120010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 704-91-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD06804571
InChI Key DNCVTVVLMRHJCJ-UHFFFAOYSA-N
Synonym indazole-6-carboxylic acid, 1h-indazole-6-carboxylicacid, 6-carboxyindazole, 6-carboxy-1h-indazole, 6-indazolecarboxylic acid, ksc376q7p, 1h-indazole-6-carboxylic acid, 6-1h indazole carboxylic acidShow MoreShow Less
PubChem CID 16227938
IUPAC Name 1H-indazole-6-carboxylic acid
SMILES OC(=O)C1=CC=C2C=NNC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 297.0°C to 300.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazole-6-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Indazole-6-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-Indazole-6-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.