missing translation for 'onlineSavingsMsg'
Learn More
1H-Indole-3-acetic acid, 98%, Thermo Scientific Chemicals Product Code.: 10275350

1H-Indole-3-acetic acid, 98%, Thermo Scientific Chemicals

Product Code. 10275350
10 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10275350

Brand: Thermo Scientific Acros 122160100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1H-Indole-3-acetic acid, CAS # 87-51-4, also known as indole-3-acetic acid, is a monocarboxylic acid mainly used as a plant growth hormone and is the most common naturally occurring plant hormone of the auxin family.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• 1H-Indole-3-acetic acid is a plant growth hormone that belongs to the auxin class. It is also considered to be an inducer of plant cell elongation and division that can cause uncontrolled growth

Applications

• This compound is a monocarboxylic acid; an acetic acid in which one of the methyl hydrogens has been replaced by a 1H-indol-3-yl group.
• Beyond its role as a plant hormone, this compound regulates tissue differentiation, apical dominance, and responses to light, gravity, and pathogens
• It can be used as a signaling molecule necessary for development of plant organs and coordination of growth

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-51-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005636
InChI Key SEOVTRFCIGRIMH-UHFFFAOYSA-N
Synonym indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon aShow MoreShow Less
PubChem CID 802
ChEBI CHEBI:16411
IUPAC Name 2-(1H-indol-3-yl)acetic acid
SMILES OC(=O)CC1=CNC2=CC=CC=C12

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 167.0°C
Color Colorless to Yellow
Flash Point 171°C
Infrared Spectrum Authentic
Assay Percent Range 98% (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 22, 66
Merck Index 15, 5004
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: soluble in acetone and ether,freely soluble in alcohol,sparingly soluble in chloroform
Formula Weight 175.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1H-Indole-3-acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-(1H-indol-3-yl)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-Indole-3-acetic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.