missing translation for 'onlineSavingsMsg'
Learn More
1H-Pyrazole-1-carboxamidine hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 15417118

1H-Pyrazole-1-carboxamidine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 15417118
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 15417118

Brand: Thermo Scientific Alfa Aesar H60631.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies:

  • Preparation of guanidylated hollow fiber membranes.
  • SGuanylation of amines and in peptide synthesis.
  • Synthesis of bis-guanidinium-cholesterol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies: • Preparation of guanidylated hollow fiber membranes. • Guanylation of amines and in peptide synthesis. • Synthesis of bis-guanidinium-cholesterol derivatives.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4023-02-3
MDL Number MFCD00210087
IUPAC Name pyrazole-1-carboximidamide;hydrochloride

Specifications

Melting Point 167°C to 170°C
Molecular Formula C4H6N4
Quantity 50 g
Synonym 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl
Solubility Information Soluble in water.
InChI Key UCQFSGCWHRTMGG-UHFFFAOYSA-N
SMILES C1=CN(N=C1)C(=N)N.Cl
Molecular Weight (g/mol) 110.12
PubChem CID 2734672
Formula Weight 146.58
Percent Purity 99%
Chemical Name or Material 1H-Pyrazole-1-carboxamidine hydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1H-Pyrazole-1-carboxamidine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Pyrazole-1-carboxamidine monohydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-Pyrazole-1-carboxamidine hydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.