missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11441140
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37718-11-9
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00011558
InChI Key IMBBXSASDSZJSX-UHFFFAOYSA-N
Synonym 4-pyrazolecarboxylic acid, 4-carboxypyrazole, pyrazole-4-carboxylic acid, unii-4xjz84gv8e, 1h-pyrazole-4-carboxylicacid, 4-carboxy-1h-pyrazole, 4xjz84gv8e, pyrazole-4-carboxylic, pubchem13293, pubchem22749
PubChem CID 3015937
IUPAC Name 1H-pyrazole-4-carboxylic acid
SMILES C1=C(C=NN1)C(=O)O

Specifications

Melting Point 281°C to 283°C
Quantity 5 g
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material 1H-Pyrazole-4-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.