missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-N,N-Bis(2-(diphenylphosphino)benzyl)cyclohexane-1,2-diamine, 95%, Thermo Scientific™

Product Code. 10656840
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 174758-63-5
CAS Min % 94.0
CAS Max % 100.0
Melting Point >300.0°C
Color Yellow
Elemental Analysis 74.96 to 84.52% C, 3.98 to 4.48% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C44H44N2P2
MDL Number MFCD16618374
Quantity 250 mg
Synonym 1r,2r-n1,n2-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1r,2r-n,n-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1r,2r-n,n'-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1r,2r-n1,n2-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine, 1r,2r-1,2-bis 2-diphenylphosphino benzylamino cyclohexane, 1r,2r-1-n,2-n-bis 2-diphenylphosphanylphenyl methyl cyclo, trans-n,n'-bis 2-diphenylphosphanylbenzyl cyclohexane-1,2-diamine, 1r,2r-n1,n2-bis 2-diphenylphosphanyl benzyl cyclohexane-1,2-diamine
InChI Key OBHPYVNBXWUKNY-NCRNUEESSA-N
SMILES C1CCC(C(C1)NCC2=CC=CC=C2P(C3=CC=CC=C3)C4=CC=CC=C4)NCC5=CC=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7
IUPAC Name (1R,2R)-1-N,2-N-bis[(2-diphenylphosphanylphenyl)methyl]cyclohexane-1,2-diamine
Molecular Weight (g/mol) 662.79
PubChem CID 10909155
Formula Weight 662.79
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-N,N-Bis(2-(Diphenylphosphino)benzyl)cyclohexane-1,2-diamine
Show More Show Less
compliance-icons
Product Identifier
  • (1R,2R)-N,N-Bis(2-(diphenylphosphino)benzyl)cyclohexane-1,2-diamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'