missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine, 95%, Thermo Scientific Chemicals

269.34 € - 1002.99 €

Chemical Identifiers

CAS 68737-65-5
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00671527,MFCD03001702
InChI Key JRHPOFJADXHYBR-HTQZYQBOSA-P
Synonym 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine, 1r,2r---n,n'-dimethylcyclohexane-1,2-diamine, trans-n1,n2-dimethylcyclohexane-1,2-diamine, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine, trans-n,n'-dimethylcyclohexane-1,2-diamine, trans-n,n'-dimethyl-1,2-cyclohexanediamine, trans-1,2-bis methylamino cyclohexane, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine, 1r,2r---1,2-bis methylamino cyclohexane
PubChem CID 2733821
IUPAC Name (1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine
SMILES C[NH2+][C@@H]1CCCC[C@H]1[NH2+]C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10511171
View Documents
Thermo Scientific Acros
337830010
1 g Glass bottle
269.34 €
1g
Please to purchase this item. Need a web account? Register with us today!
10490364
View Documents
Thermo Scientific Acros
337830050
5 g Glass bottle
1002.99 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

68737-65-5
144.26
JRHPOFJADXHYBR-HTQZYQBOSA-P
2733821
C[NH2+][C@@H]1CCCC[C@H]1[NH2+]C
C8H20N2
MFCD00671527,MFCD03001702
1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine, 1r,2r---n,n'-dimethylcyclohexane-1,2-diamine, trans-n1,n2-dimethylcyclohexane-1,2-diamine, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine, trans-n,n'-dimethylcyclohexane-1,2-diamine, trans-n,n'-dimethyl-1,2-cyclohexanediamine, trans-1,2-bis methylamino cyclohexane, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine, 1r,2r---1,2-bis methylamino cyclohexane
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine

Specifications

68737-65-5
100.0
Authentic
C8H20N2
MFCD00671527,MFCD03001702
1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine, 1r,2r---n,n'-dimethylcyclohexane-1,2-diamine, trans-n1,n2-dimethylcyclohexane-1,2-diamine, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine, trans-n,n'-dimethylcyclohexane-1,2-diamine, trans-n,n'-dimethyl-1,2-cyclohexanediamine, trans-1,2-bis methylamino cyclohexane, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine, 1r,2r---1,2-bis methylamino cyclohexane
C[NH2+][C@@H]1CCCC[C@H]1[NH2+]C
144.26
142.24
Low Melting Solid
94.0
White to Yellow
94% min. (GC)
C6H10(NHCH3)2
− 143.00
JRHPOFJADXHYBR-HTQZYQBOSA-P
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine
2733821
95%
(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.