missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%, Thermo Scientific Chemicals

137.00 € - 1436.00 €

Chemical Identifiers

CAS 79200-57-0
Molecular Formula C6H13NO3·HCl
Molecular Weight (g/mol) 183.63
InChI Key BLTXEPQZAMUGID-ILLHIODVSA-N
Synonym 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, n-4-mesityl-1,3-thiazol-2-yl benzamide, 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hcl, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-am, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl cyclopentanaminium chloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxylmethyl-1-aminocyclopentan hydro-chloride, 1r,2s,3r,5r-3-amino-5-hydroxymethyl-cyclopentane-1,2-diol hydrochloride
PubChem CID 2734536
IUPAC Name (1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride
SMILES C1C(C(C(C1N)O)O)CO.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10058891
View Documents Promotion Details
Thermo Scientific Acros
362522500
250 mg Glass bottle
137.00 €
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10540732
View Documents Promotion Details
Thermo Scientific Acros
362520010
1 g Glass bottle
364.00 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11984641
View Documents Promotion Details
Thermo Scientific Acros
362520050
5 g Glass bottle
1436.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

79200-57-0
183.63
1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, n-4-mesityl-1,3-thiazol-2-yl benzamide, 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hcl, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-am, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl cyclopentanaminium chloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxylmethyl-1-aminocyclopentan hydro-chloride, 1r,2s,3r,5r-3-amino-5-hydroxymethyl-cyclopentane-1,2-diol hydrochloride
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride
C6H13NO3·HCl
BLTXEPQZAMUGID-ILLHIODVSA-N
2734536
C1C(C(C(C1N)O)O)CO.Cl

Specifications

79200-57-0
C6H13NO3·HCl
BLTXEPQZAMUGID-ILLHIODVSA-N
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol;hydrochloride
2734536
95%
94% min. (HPLC)
1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hydrochloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-aminocyclopentane hydrochloride, n-4-mesityl-1,3-thiazol-2-yl benzamide, 1r,2s,3r,5r-3-amino-5-hydroxymethyl cyclopentane-1,2-diol hcl, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl-1-am, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxymethyl cyclopentanaminium chloride, 1r,2s,3r,4r-2,3-dihydroxy-4-hydroxylmethyl-1-aminocyclopentan hydro-chloride, 1r,2s,3r,5r-3-amino-5-hydroxymethyl-cyclopentane-1,2-diol hydrochloride
C1C(C(C(C1N)O)O)CO.Cl
183.63
183.63
(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • (1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only