missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(+)-Camphoric acid, 98+%, Thermo Scientific Chemicals

30.05 € - 271.35 €

Chemical Identifiers

CAS 124-83-4
Molecular Formula C10H14O4-2
Molecular Weight (g/mol) 198.218
MDL Number MFCD00001375
InChI Key LSPHULWDVZXLIL-LDWIPMOCSA-L
Synonym d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
PubChem CID 6918944
IUPAC Name (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
SMILES CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11446316
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23106.14
25 g
30.05 €
25g
Please to purchase this item. Need a web account? Register with us today!
11456316
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23106.22
100 g
76.18 €
100g
Please to purchase this item. Need a web account? Register with us today!
11466316
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23106.36
500 g
271.35 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-83-4
198.218
LSPHULWDVZXLIL-LDWIPMOCSA-L
6918944
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C
C10H14O4-2
MFCD00001375
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate

Specifications

124-83-4
C10H14O4-2
2050204
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C
198.218
200.23
+46° (c=1 in Methanol)
183°C to 188°C
MFCD00001375
141733
LSPHULWDVZXLIL-LDWIPMOCSA-L
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
6918944
≥98%
(1R,3S)-(+)-Camphoric acid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-715-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only